siRNA Details
Gene Symbol: Gpr161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081126.1 | NP_001074595.1 | 4 | 763 | ||
GenBank mRNA | AK145635.1 | - | 3 | 787 | ||
AY255596.1 | - | 2 | 254 | |||
BC137659.1 | - | 3 | 701 | 8860678 | ||
EF197953.1 | - | 3 | 788 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
