siRNA Details
Gene Symbol: Ralgps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290570.1 | NP_001277499.1 | 5 | 558 | ||
NM_001290572.1 | NP_001277501.1 | 6 | 609 | |||
NM_175211.5 | NP_780420.1 | 6 | 609 | |||
GenBank mRNA | AK076767.1 | - | 6 | 594 | ||
AK129123.1 | - | 5 | 525 | |||
AK170248.1 | - | 6 | 556 | |||
BC072656.1 | - | 6 | 544 | 30536315 |

More Information
Other Species Targeted

Gene Ontology Categories:
