siRNA Details
Gene Symbol: Zfp385b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113399.1 | NP_001106870.1 | 3 | 452 | ||
NM_001113400.1 | NP_001106871.1 | 3 | 474 | |||
NM_178723.6 | NP_848838.2 | 5 | 984 | |||
GenBank mRNA | AK046798.1 | - | 5 | 985 | ||
AK050293.1 | - | 3 | 475 | |||
AK078551.1 | - | 3 | 427 | |||
AK079322.1 | - | 5 | 867 | |||
AK149352.1 | - | 5 | 965 | |||
BC048543.1 | - | 3 | 452 | |||
BC132352.1 | - | 3 | 563 | 40130629 | ||
BC138239.1 | - | 3 | 563 | 8861258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
