siRNA Details
Gene Symbol: Fam171b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175514.2 | NP_780723.1 | 3 | 558 | ||
GenBank mRNA | AK038899.1 | - | 3 | 651 | ||
AK052508.1 | - | 3 | 559 | |||
BC113787.1 | - | 3 | 571 | 40060749 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
