siRNA Details
Gene Symbol: Tspan18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183180.2 | NP_899003.1 | 7 | 784 | ||
GenBank mRNA | AK032765.1 | - | 6 | 695 | ||
BC052169.1 | - | 7 | 747 | 6335221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
