siRNA Details
Gene Symbol: Lrrc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081358.2 | NP_001074827.1 | 22 | 4428 | ||
NM_001291452.1 | NP_001278381.1 | 22 | 4428 | |||
NM_001291453.1 | NP_001278382.1 | 22 | 4428 | |||
GenBank mRNA | AK122498.1 | - | 22 | 3563 | ||
BC138450.1 | - | 22 | 4335 | 8861470 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
