siRNA Details
Gene Symbol: Atp6v0d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175406.3 | NP_780615.2 | 8 | 1071 | ||
GenBank mRNA | AB088358.1 | - | 8 | 1027 | ||
AK075745.1 | - | 8 | 1111 | |||
AK089507.1 | - | 8 | 996 | |||
AK137586.1 | - | 8 | 1053 | |||
AK170072.1 | - | 8 | 1045 | |||
AK170405.1 | - | 8 | 1044 | |||
AK170578.1 | - | 8 | 1015 | |||
AK170659.1 | - | 8 | 1020 | |||
AY145896.1 | - | 8 | 1071 | |||
AY517482.1 | - | 8 | 1063 | |||
BC087899.1 | - | 8 | 1005 | 30504725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
