siRNA Details
Gene Symbol: Tmem245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175518.5 | NP_780727.3 | 9 | 1589 | ||
GenBank mRNA | AK052826.1 | - | 9 | 1584 |

More Information
Other Species Targeted

Gene Ontology Categories:
