siRNA Details
Gene Symbol: Bnc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172870.4 | NP_766458.3 | 5 | 1300 | ||
GenBank mRNA | AK030323.1 | - | 5 | 1300 | ||
AY438375.1 | - | 5 | 1275 | |||
BC110483.2 | - | 4 | 1180 | 40048720 | ||
BC110484.1 | - | 4 | 1180 | 40048722 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
