siRNA Details
Gene Symbol: Skint5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167876.1 | NP_001161348.1 | 2 | 80 | ||
NM_001167878.1 | NP_001161350.1 | 2 | 80 | |||
GenBank mRNA | EU099301.1 | - | 2 | 80 | ||
EU099302.1 | - | 2 | 80 | |||
EU099303.1 | - | 2 | 80 |
