siRNA Details
Gene Symbol: Lactbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243262.1 | NP_001230191.1 | 8 | 1584 | ||
GenBank mRNA | BC029684.1 | - | 8 | 3072 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
