siRNA Details
Gene Symbol: Lrrc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162983.1 | NP_001156455.1 | 2 | 1175 | ||
GenBank mRNA | AK038658.1 | - | 2 | 1139 | ||
AK045936.1 | - | 2 | 727 | |||
AK078190.1 | - | 2 | 1150 | |||
AK160116.1 | - | 2 | 265 | |||
BC129963.1 | - | 2 | 1168 | |||
BC129964.1 | - | 3 | 1098 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
