siRNA Details
Gene Symbol: Cct8l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198621.2 | NP_941023.1 | 1 | 896 | ||
GenBank mRNA | AK133375.1 | - | 1 | 896 | ||
BC050797.1 | - | 1 | 813 | 6743432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
