siRNA Details
Gene Symbol: Gprin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183183.2 | NP_899006.1 | 2 | 2353 | ||
GenBank mRNA | AK050148.1 | - | 2 | 2354 | ||
AK173336.1 | - | 2 | 2357 | |||
BC138149.1 | - | 1 | 2223 | 8861168 | ||
BC138150.1 | - | 1 | 2223 | 8861169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
