siRNA Details
Gene Symbol: Igkv1-135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BF301241.1 | - | 375 | |||
M17722.1 | - | 2 | 333 | |||
M20830.1 | - | 2 | 371 | |||
M91696.1 | - | 1 | 299 | |||
U30241.1 | - | 1 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
