siRNA Details
Gene Symbol: Mrgprb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175531.4 | NP_780740.2 | 2 | 529 | ||
GenBank mRNA | AK029369.1 | - | 2 | 530 | ||
BC107390.1 | - | 2 | 527 | 40057815 | ||
BC107391.2 | - | 2 | 527 | 40057816 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
