siRNA Details
Gene Symbol: Rgma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177740.5 | NP_808408.2 | 4 | 1508 | ||
GenBank mRNA | AJ557513.1 | - | 3 | 1262 | ||
AK082560.1 | - | 3 | 1405 | |||
AK145224.1 | - | 3 | 1253 | |||
BC023870.1 | - | 3 | 1316 | 5353958 | ||
BC059072.1 | - | 4 | 1508 | 6826181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
