siRNA Details
Gene Symbol: Dlgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145965.1 | NP_001139437.1 | 14 | 3182 | ||
NM_172910.3 | NP_766498.2 | 15 | 3281 | |||
GenBank mRNA | AK032564.1 | - | 15 | 3282 | ||
AK033077.1 | - | 7 | 1332 | |||
AY243847.3 | - | 9 | 2651 | |||
BC118969.2 | - | 14 | 2968 | 40097764 |

More Information
Other Species Targeted

Gene Ontology Categories:
