siRNA Details
Gene Symbol: Dlgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145965.1 | NP_001139437.1 | 5 | 618 | ||
NM_172910.3 | NP_766498.2 | 6 | 717 | |||
GenBank mRNA | AK032564.1 | - | 6 | 718 | ||
AK082945.1 | - | 4 | 490 | |||
AY243847.3 | - | 1 | 129 | |||
BC118969.2 | - | 5 | 404 | 40097764 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
