siRNA Details
Gene Symbol: Defb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153108.4 | NP_694748.3 | 2 | 146 | ||
GenBank mRNA | AJ300674.1 | - | 2 | 146 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
