siRNA Details
Gene Symbol: Nanos3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194059.2 | NP_918948.1 | 1 | 373 | ||
GenBank mRNA | AB095973.1 | - | 1 | 373 | ||
BC119513.1 | - | 1 | 373 | 40055381 | ||
BC119514.1 | - | 1 | 373 | 40055383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
