siRNA Details
Gene Symbol: Ccdc113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172914.2 | NP_766502.1 | 2 | 213 | ||
GenBank mRNA | AK077130.1 | - | 2 | 214 | ||
BC048506.1 | - | 2 | 174 | |||
BC060957.1 | - | 2 | 211 | 6705542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
