siRNA Details
Gene Symbol: Sipa1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081337.1 | NP_001074806.1 | 1 | 762 | ||
GenBank mRNA | BC072593.1 | - | 3 | 1048 | ||
BC150682.1 | - | 1 | 807 | 9087593 |

More Information
Other Species Targeted

Gene Ontology Categories:
