siRNA Details
Gene Symbol: Sipa1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081337.1 | NP_001074806.1 | 1 | 209 | ||
GenBank mRNA | BC072593.1 | - | 3 | 495 | ||
BC150682.1 | - | 1 | 254 | 9087593 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
