siRNA Details
Gene Symbol: Tnfaip8l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033535.3 | NP_001028707.1 | 2 | 703 | ||
GenBank mRNA | AK036953.1 | - | 2 | 660 | ||
AK154109.1 | - | 673 | ||||
AK170374.1 | - | 2 | 681 | |||
AK171174.1 | - | 2 | 700 | |||
BC118003.1 | - | 2 | 693 | 40057108 | ||
BC119769.1 | - | 2 | 690 | 40057107 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
