siRNA Details
Gene Symbol: Polr2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145632.2 | NP_663607.1 | 4 | 376 | ||
GenBank mRNA | AK146429.1 | - | 4 | 401 | ||
AK161617.1 | - | 4 | 397 | |||
BC002306.1 | - | 4 | 376 | 3591968 | ||
BC092087.1 | - | 4 | 286 | 5042770 | ||
BC125612.1 | - | 4 | 360 | 40130127 | ||
BC125638.1 | - | 4 | 360 | 40130153 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
