siRNA Details
Gene Symbol: Defb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139225.1 | NP_631971.1 | 2 | 159 | ||
GenBank mRNA | AJ437645.1 | - | 2 | 159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
