siRNA Details
Gene Symbol: Otoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139310.1 | NP_647471.1 | 21 | 2307 | ||
GenBank mRNA | AK157904.1 | - | 21 | 2339 | ||
AK158280.1 | - | 11 | 1280 | |||
AK158326.1 | - | 11 | 1276 | |||
AY055122.1 | - | 21 | 2307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
