siRNA Details
Gene Symbol: Csad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144942.4 | NP_659191.1 | 12 | 1190 | ||
GenBank mRNA | AK005015.1 | - | 12 | 1191 | ||
AY033912.1 | - | 12 | 1107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
