siRNA Details
Gene Symbol: Obox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145710.2 | NP_663756.2 | 2 | 139 | ||
GenBank mRNA | BY731792.1 | - | 2 | 138 | ||
CN712286.1 | - | 2 | 139 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
