siRNA Details
Gene Symbol: Vmn1r189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145844.1 | NP_665843.1 | 1 | 868 | ||
GenBank mRNA | BC150715.1 | - | 1 | 967 | 9087626 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
