siRNA Details
Gene Symbol: Olfr605 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011854.2 | NP_001011854.2 | 1 | 780 | ||
GenBank mRNA | BC147216.1 | - | 1 | 780 | 9056257 | |
BC147217.1 | - | 1 | 780 | 9056245 |

More Information
Other Species Targeted

Gene Ontology Categories:
