siRNA Details
Gene Symbol: Olfr1274-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146263.2 | NP_666375.2 | 1 | 759 | ||
GenBank mRNA | BC145937.1 | - | 759 | 40131240 |
Gene Symbol: Gm13769 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270423.1 | NP_001257352.1 | 2 | 890 |
Gene Symbol: Olfr1506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146265.2 | NP_666377.2 | 2 | 890 | ||
GenBank mRNA | BC111438.1 | - | 3 | 369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
