siRNA Details
Gene Symbol: Olfr720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146392.1 | NP_666504.1 | 1 | 53 | ||
GenBank mRNA | BC120861.1 | - | 1 | 90 | 40129784 | |
BC132043.1 | - | 1 | 90 | 40130320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
