siRNA Details
Gene Symbol: Olfr1328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146399.2 | NP_666511.2 | 1 | 626 | ||
GenBank mRNA | BC106978.2 | - | 1 | 626 | 40042180 | |
BC148290.2 | - | 1 | 626 | 40042169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
