siRNA Details
Gene Symbol: Olfr97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146512.1 | NP_666723.1 | 1 | 741 | ||
GenBank mRNA | BC134363.1 | - | 745 | 40134689 |
Gene Symbol: Olfr107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146511.2 | NP_666722.2 | 1 | 811 | ||
GenBank mRNA | BC141327.1 | - | 1 | 811 | 9055939 | |
BC141329.1 | - | 1 | 811 | 9055941 | ||
BC148321.1 | - | 1 | 738 | 40054936 |

More Information
Other Species Targeted

Gene Ontology Categories:
