siRNA Details
Gene Symbol: Olfr1501 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146633.2 | NP_666844.1 | 1 | 136 |
Gene Symbol: Olfr1504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146634.1 | NP_666845.1 | 1 | 136 | ||
GenBank mRNA | BC137835.1 | - | 1 | 233 | 8860854 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
