siRNA Details
Gene Symbol: Olfr1163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146644.2 | NP_666855.1 | 1 | 533 | ||
GenBank mRNA | BC120531.1 | - | 1 | 533 | 40129454 | |
BC120557.1 | - | 1 | 533 | 40129480 |
Gene Symbol: Olfr1162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011835.1 | NP_001011835.1 | 1 | 506 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
