siRNA Details
Gene Symbol: Olfr1446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146704.1 | NP_666915.1 | 1 | 572 | ||
GenBank mRNA | BC119061.1 | - | 1 | 655 | 8733810 | |
BC137821.1 | - | 1 | 655 | 8860840 |
Gene Symbol: Olfr1447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146703.1 | NP_666914.1 | 1 | 572 | ||
GenBank mRNA | BC125359.1 | - | 1 | 655 | 40129874 | |
BC132255.1 | - | 1 | 655 | 40130532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
