siRNA Details
Gene Symbol: Olfr429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146722.2 | NP_666933.2 | 1 | 298 | ||
GenBank mRNA | BC127973.1 | - | 1 | 298 | 40135130 |

More Information
Other Species Targeted

Gene Ontology Categories:
