siRNA Details
Gene Symbol: Olfr341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146950.1 | NP_667161.1 | 1 | 81 | ||
GenBank mRNA | BC130244.1 | - | 1 | 85 | 40142305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
