siRNA Details
Gene Symbol: Olfr181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146999.2 | NP_667210.2 | 2 | 955 | ||
GenBank mRNA | BC125397.1 | - | 1 | 668 | 40129912 | |
BC131916.1 | - | 1 | 668 | 40130193 |
Gene Symbol: Olfr178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146997.2 | NP_667208.2 | 1 | 583 | ||
GenBank mRNA | BC111099.1 | - | 1 | 582 | ||
BC127221.1 | - | 1 | 581 |
Gene Symbol: Olfr180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011662.2 | NP_001011662.2 | 2 | 765 |

More Information
Other Species Targeted

Gene Ontology Categories:
