siRNA Details
Gene Symbol: Olfr644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147121.1 | NP_667332.1 | 1 | 478 | ||
GenBank mRNA | BC099886.1 | - | 1 | 470 | ||
BC099887.1 | - | 1 | 470 |
Gene Symbol: Olfr643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147077.1 | NP_667288.1 | 1 | 478 | ||
GenBank mRNA | BC106803.2 | - | 1 | 478 | 40042567 | |
BC146607.1 | - | 1 | 478 | 40042594 | ||
BC148182.1 | - | 1 | 478 | 40043246 |

More Information
Other Species Targeted

Gene Ontology Categories:
