siRNA Details
Gene Symbol: Cetn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007593.5 | NP_031619.3 | 1 | 195 | ||
GenBank mRNA | AK005736.1 | - | 1 | 174 | ||
AK005743.1 | - | 1 | 156 | |||
AK005837.1 | - | 1 | 196 | |||
AK006940.1 | - | 1 | 196 | |||
AK015808.1 | - | 194 | ||||
AK076986.1 | - | 205 | ||||
AK132753.1 | - | 1 | 195 | |||
BC048488.1 | - | 1 | 196 | 6775067 | ||
BC061155.1 | - | 1 | 194 | 6774850 | ||
BU961997.1 | - | 1 | 233 | |||
D16301.1 | - | 1 | 179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
