siRNA Details
Gene Symbol: Map3k12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163643.1 | NP_001157115.1 | 15 | 2760 | ||
NM_009582.4 | NP_033608.3 | 13 | 2584 | |||
GenBank mRNA | AK029882.1 | - | 13 | 2393 | ||
AK034374.1 | - | 15 | 2576 | |||
AK081623.1 | - | 8 | 1252 | |||
AK166435.1 | - | 15 | 2680 | |||
BC047158.1 | - | 12 | 2378 | 6494216 | ||
BC057572.1 | - | 15 | 2584 | 6401321 | ||
U14636.1 | - | 13 | 2416 | |||
U23789.1 | - | 12 | 2374 |

More Information
Other Species Targeted

Gene Ontology Categories:
