siRNA Details
Gene Symbol: Prl2c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011954.2 | NP_036084.2 | 170 | |||
GenBank mRNA | BC058816.1 | - | 170 | 30053850 | ||
BC064772.1 | - | 161 | 30434146 | |||
BC100299.1 | - | 152 | 30713114 |
Gene Symbol: Prl2c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031191.1 | NP_112468.1 | 2 | 117 | ||
GenBank mRNA | BC056203.1 | - | 2 | 167 | 30014401 | |
BC064771.1 | - | 2 | 173 | 30049431 | ||
BC080826.1 | - | 2 | 182 | 30438424 | ||
BC100300.1 | - | 2 | 152 | 30715342 | ||
BC145439.1 | - | 2 | 145 | 9053997 | ||
K02245.1 | - | 2 | 117 | |||
X75557.1 | - | 2 | 121 |
Gene Symbol: Prl2c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011118.2 | NP_035248.2 | 2 | 180 | ||
GenBank mRNA | AK145995.1 | - | 2 | 182 | ||
AK164068.1 | - | 2 | 181 | |||
BC132078.1 | - | 2 | 162 | 40130355 | ||
BC132080.1 | - | 2 | 162 | 40130357 | ||
K03235.1 | - | 2 | 179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
