siRNA Details
Gene Symbol: Pask | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080850.2 | NP_543126.2 | 10 | 2212 | ||
GenBank mRNA | AK028435.1 | - | 10 | 2214 | ||
AK083528.1 | - | 10 | 2183 | |||
AK083590.1 | - | 10 | 2177 | |||
AK129063.1 | - | 10 | 2102 | |||
AK172031.1 | - | 10 | 2160 | |||
BC150821.1 | - | 9 | 1924 | 9087732 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
