siRNA Details
Gene Symbol: Acvr1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033369.3 | NP_001028541.1 | 3 | 300 | ||
NM_001111030.1 | NP_001104500.1 | 4 | 667 | |||
GenBank mRNA | AK049089.1 | - | 3 | 301 | ||
AK142396.1 | - | 4 | 670 | |||
BC145224.1 | - | 3 | 463 |

More Information
Other Species Targeted

Gene Ontology Categories:
