siRNA Details
Gene Symbol: Smok4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029728.1 | - | 2 | 977 | ||
BC125398.1 | - | 1 | 535 | |||
BC132092.1 | - | 1 | 535 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
