siRNA Details
Gene Symbol: Slc25a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013770.2 | NP_038798.2 | 4, 5 | 532 | ||
GenBank mRNA | AF188712.1 | - | 4, 5 | 503 | ||
AK008038.1 | - | 4, 5 | 502 | |||
AK050282.1 | - | 4, 5 | 486 | |||
AK083831.1 | - | 4, 5 | 510 | |||
AK151645.1 | - | 4, 5 | 532 | |||
BC003222.1 | - | 4, 5 | 450 | 3482316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
